ENA429358949 Enamine

2-chloro-6-(pyrrolidin-1-yl)-9H-purine

  • MDL number MFCD22375190
  • Cas Number : 897936-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186662
Availability details for
Quantity
  • Availability
  • Add to Cart