ENA429358950 Enamine

1-[3-(dimethylamino)propyl]-6-hydroxy-4-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD09847910
  • Cas Number : 40859-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186663

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186663
Availability details for
Quantity
  • Availability
  • Add to Cart