ENA429358951 Enamine

3-(1-methyl-1,2,3,6-tetrahydropyridin-4-yl)-1H-indole

  • MDL number MFCD11840338
  • Cas Number : 17403-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186664
Availability details for
Quantity
  • Availability
  • Add to Cart