ENA429358952 Enamine

3-{6-methoxy-[1,2,4]triazolo[4,3-b]pyridazin-3-yl}propanoic acid

  • MDL number MFCD19691808
  • Cas Number : 1322604-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186665
Availability details for
Quantity
  • Availability
  • Add to Cart