ENA429358957 Enamine

ethyl 4-hydroxy-2-oxo-1,2-dihydroquinoline-3-carboxylate

  • MDL number MFCD00218345
  • Cas Number : 40059-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186670
Availability details for
Quantity
  • Availability
  • Add to Cart