ENA429358964 Enamine

5-(propan-2-yl)-2-(1H-pyrrol-1-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD19691831
  • Cas Number : 1322604-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186678

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186678
Availability details for
Quantity
  • Availability
  • Add to Cart