ENA429358965 Enamine

ethyl 4-chloro-2-oxo-1,2-dihydroquinoline-3-carboxylate

  • MDL number MFCD00572655
  • Cas Number : 99429-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-203
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186679
Availability details for
Quantity
  • Availability
  • Add to Cart