ENA429358968 Enamine

7-bromo-[1,3]oxazolo[4,5-b]pyridine

  • MDL number MFCD24038545
  • Cas Number : 1429901-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186683

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-186683
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart