ENA429358974 Enamine

2-(3-oxo-2,3-dihydro-1H-indazol-2-yl)acetic acid

  • MDL number MFCD16171059
  • Cas Number : 64697-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186689

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186689
Availability details for
Quantity
  • Availability
  • Add to Cart