ENA429358979 Enamine

3-[(4-methyl-1,3-thiazol-2-yl)carbamoyl]propanoic acid

  • MDL number MFCD01080037
  • Cas Number : 313660-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186694
Availability details for
Quantity
  • Availability
  • Add to Cart