ENA429358980 Enamine

3-[(dimethyl-1,3-thiazol-2-yl)carbamoyl]propanoic acid

  • MDL number MFCD00971902
  • Cas Number : 304459-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186695

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186695
Availability details for
Quantity
  • Availability
  • Add to Cart