ENA429358990 Enamine

methyl 3-amino-4-fluoro-1-benzothiophene-2-carboxylate

  • MDL number MFCD02091511
  • Cas Number : 144899-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186706
Availability details for
Quantity
  • Availability
  • Add to Cart