ENA429358992 Enamine

2-[4-(1H-1,2,3,4-tetrazol-1-yl)thian-4-yl]acetic acid

  • MDL number MFCD22201612
  • Cas Number : 1374407-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186708
Availability details for
Quantity
  • Availability
  • Add to Cart