ENA429358999 Enamine

2-methoxy-5-(1H-pyrrol-1-yl)aniline

  • MDL number MFCD13807250
  • Cas Number : 946699-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186716

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186716
Availability details for
Quantity
  • Availability
  • Add to Cart