ENA429359003 Enamine

2-[4-(1H-pyrrol-1-yl)thian-4-yl]acetic acid

  • MDL number MFCD26523040
  • Cas Number : 1573547-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-186720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186720
Availability details for
Quantity
  • Availability
  • Add to Cart