ENA429359005 Enamine

2-(1-amino-4-tert-butylcyclohexyl)acetic acid

  • MDL number MFCD20384964
  • Cas Number : 1339351-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186722
Availability details for
Quantity
  • Availability
  • Add to Cart