ENA429359007 Enamine

2-[1-(1H-pyrrol-1-yl)cyclohexyl]acetic acid

  • MDL number MFCD20386918
  • Cas Number : 1343341-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186724

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186724
Availability details for
Quantity
  • Availability
  • Add to Cart