ENA429359008 Enamine

3-phenyl-3-(1H-pyrrol-1-yl)propanoic acid

  • MDL number MFCD03768199
  • Cas Number : 75391-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186725
Availability details for
Quantity
  • Availability
  • Add to Cart