ENA429359010 Enamine

7-chloro-4-hydroxyquinoline-3-carboxylic acid

  • MDL number MFCD00038080
  • Cas Number : 86-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186727

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186727
Availability details for
Quantity
  • Availability
  • Add to Cart