ENA429359020 Enamine

N-(2-aminoethyl)-1-methyl-1H-indole-2-carboxamide

  • MDL number MFCD11868352
  • Cas Number : 1146293-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186737
Availability details for
Quantity
  • Availability
  • Add to Cart