ENA429359022 Enamine

1-ethyl-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD06447022
  • Cas Number : 24667-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186739

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186739
Availability details for
Quantity
  • Availability
  • Add to Cart