ENA429359033 Enamine

2,7-diazatricyclo[6.4.0.0^{2,6}]dodeca-1(8),6,9,11-tetraene

  • MDL number MFCD01117053
  • Cas Number : 7724-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186750

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186750
Availability details for
Quantity
  • Availability
  • Add to Cart