ENA429359037 Enamine

2-(pyridin-4-yl)-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD00453950
  • Cas Number : 1724-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-88
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186754
Availability details for
Quantity
  • Availability
  • Add to Cart