ENA429359041 Enamine

2-(pyridin-3-yl)-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD00453951
  • Cas Number : 1571-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186758
Availability details for
Quantity
  • Availability
  • Add to Cart