ENA429359046 Enamine

1-(4-fluorophenyl)-4-(4-methylpiperidin-1-yl)butan-1-one

  • MDL number MFCD00867737
  • Cas Number : 3575-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186797

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186797
Availability details for
Quantity
  • Availability
  • Add to Cart