ENA429359047 Enamine

5-bromo-3-{[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]methyl}-3,4-dihydropyrimidin-4-one

  • MDL number MFCD22314714
  • Cas Number : 1510711-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186821
Availability details for
Quantity
  • Availability
  • Add to Cart