ENA429359054 Enamine

(5-methyl-1,2-thiazol-3-yl)methanol

  • MDL number MFCD26954838
  • Cas Number : 1803598-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186866
Availability details for
Quantity
  • Availability
  • Add to Cart