ENA429359059 Enamine

methyl 2-hydroxy-2-[3-(trifluoromethoxy)phenyl]acetate

  • MDL number MFCD21085066
  • Cas Number : 1480321-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186925
Availability details for
Quantity
  • Availability
  • Add to Cart