ENA429359060 Enamine

2-hydroxy-2-[3-(trifluoromethoxy)phenyl]acetic acid

  • MDL number MFCD20868635
  • Cas Number : 1214378-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186926
Availability details for
Quantity
  • Availability
  • Add to Cart