ENA429359066 Enamine

4-(3,5-dichlorophenoxy)pyrimidin-2-amine

  • MDL number MFCD23995274
  • Cas Number : 1503974-35-3
  • Molecular Weight : 256.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-186979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186979
Availability details for
Quantity
  • Availability
  • Add to Cart