ENA429359069 Enamine

3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile

  • MDL number MFCD00052315
  • Cas Number : 27328-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-186991
Availability details for
Quantity
  • Availability
  • Add to Cart