ENA429359070 Enamine

1-bromo-3-cyclopropylpropan-2-one

  • MDL number MFCD18815931
  • Cas Number : 658076-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186994
Availability details for
Quantity
  • Availability
  • Add to Cart