ENA429359071 Enamine

5-bromo-2-methyl-3-[(3-methyl-1,2,4-oxadiazol-5-yl)methyl]-3,4-dihydropyrimidin-4-one

  • MDL number MFCD22801743
  • Cas Number : 1510311-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-186998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186998
Availability details for
Quantity
  • Availability
  • Add to Cart