ENA429359072 Enamine

5-bromo-3-[(3-ethyl-1,2,4-oxadiazol-5-yl)methyl]-2-methyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD22315091
  • Cas Number : 1537304-89-4
  • Molecular Weight : 299.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-187003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187003
Availability details for
Quantity
  • Availability
  • Add to Cart