ENA429359073 Enamine

3-(6-methyl-1,2,3,4-tetrahydroquinolin-1-yl)butan-2-one

  • MDL number MFCD12147135
  • Cas Number : 1152866-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187009

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187009
Availability details for
Quantity
  • Availability
  • Add to Cart