ENA429359075 Enamine

3-bromo-5-(hydroxymethyl)benzonitrile

  • MDL number MFCD09261039
  • Cas Number : 1205515-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187016
Availability details for
Quantity
  • Availability
  • Add to Cart