ENA429359078 Enamine

2-(4-fluorophenyl)-1-(4-methylphenyl)ethan-1-one

  • MDL number MFCD03788454
  • Cas Number : 684270-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187046

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187046
Availability details for
Quantity
  • Availability
  • Add to Cart