ENA429359079 Enamine

4-(1-methyl-1H-pyrazol-4-yl)pyrimidin-2-amine

  • MDL number MFCD16766989
  • Cas Number : 1247765-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187048
Availability details for
Quantity
  • Availability
  • Add to Cart