ENA429359080 Enamine

5-chloro-1-N-phenylbenzene-1,2-diamine

  • MDL number MFCD01156582
  • Cas Number : 68406-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187049
Availability details for
Quantity
  • Availability
  • Add to Cart