ENA429359081 Enamine

2-{2-[1-(methylamino)ethyl]phenyl}-1lambda6,2-thiazolidine-1,1-dione hydrochloride

  • MDL number MFCD28118589
  • Cas Number : 1803609-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187050
Availability details for
Quantity
  • Availability
  • Add to Cart