ENA429359084 Enamine

2,6-dimethylbenzene-1,4-diamine dihydrochloride

  • MDL number MFCD18973468
  • Cas Number : 56496-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187053
Availability details for
Quantity
  • Availability
  • Add to Cart