ENA429359088 Enamine

4-(2-aminoethyl)-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD28118593
  • Cas Number : 1795184-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187059
Availability details for
Quantity
  • Availability
  • Add to Cart