ENA429359094 Enamine

1-(2-amino-6-chloropyrimidin-4-yl)piperidin-3-ol

  • MDL number MFCD23715360
  • Cas Number : 1386832-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187074

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187074
Availability details for
Quantity
  • Availability
  • Add to Cart