ENA429359096 Enamine

methyl 2-[(4-bromo-2-nitrophenyl)amino]-2-methylpropanoate

  • MDL number MFCD28118595
  • Cas Number : 1803561-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187076
Availability details for
Quantity
  • Availability
  • Add to Cart