ENA429359097 Enamine

4-(morpholin-4-yl)but-3-en-2-one

  • Cas Number : 90054-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187079
Availability details for
Quantity
  • Availability
  • Add to Cart