ENA429359101 Enamine

2-[3-(trifluoromethyl)phenyl]ethanethioamide

  • MDL number MFCD09817423
  • Cas Number : 952182-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187089
Availability details for
Quantity
  • Availability
  • Add to Cart