ENA429359102 Enamine

2-[cyclopropyl(methyl)amino]ethan-1-ol

  • MDL number MFCD12186661
  • Cas Number : 24907-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187092
Availability details for
Quantity
  • Availability
  • Add to Cart