ENA429359105 Enamine

methyl 4-bromo-2-chloro-5-sulfamoylbenzoate

  • MDL number MFCD18842212
  • Cas Number : 1343862-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187099

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187099
Availability details for
Quantity
  • Availability
  • Add to Cart