ENA429359106 Enamine

3-(4-bromophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD19328381
  • Cas Number : 21724-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187101
Availability details for
Quantity
  • Availability
  • Add to Cart