ENA429359109 Enamine

3-methyldodecanoic acid

  • MDL number MFCD02258681
  • Cas Number : 13490-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187114

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187114
Availability details for
Quantity
  • Availability
  • Add to Cart